Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026422.6 | NP_001021593.1 | 4 - 5 | 1206 | 66 | |
NM_001026425.6 | NP_001021596.1 | 8 - 9 | 1077 | 66 | ||
NM_001026427.7 | NP_001021598.2 | 9 - 10 | 1167 | 66 | ||
NM_001377627.2 | NP_001364785.1 | 7 - 8 | 1146 | 66 | ||
NM_001377628.3 | NP_001364784.1 | 7 - 8 | 1140 | 66 | ||
NM_001381205.1 | NP_001367893.1 | 9 - 10 | 1239 | 66 | ||
NM_001381206.1 | NP_001368498.1 | 9 - 10 | 1161 | 66 | ||
NM_001381207.1 | NP_001368499.1 | 8 - 9 | 1071 | 66 | ||
NM_001381208.1 | NP_001368501.1 | 6 - 7 | 837 | 66 | ||
NM_001381209.1 | NP_001368502.1 | 6 - 7 | 831 | 66 | ||
NM_001381210.1 | NP_001367892.1 | 3 - 4 | 525 | 66 | ||
GenBank mRNA | AF020342.1 | - | 7 - 8 | 1444 | 66 | |
AF020343.1 | - | 7 - 8 | 1436 | 66 | ||
AF020344.1 | - | 4 - 5 | 1255 | 66 | ||
AF032112.1 | - | 7 - 8 | 1479 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |