Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhhc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060606.6 | NP_493007.2 | 9 - 10 | 1255 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |