Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060656.4 | NP_493057.1 | 15 - 16 | 3725 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |