Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060710.2 | NP_493111.1 | 9 - 10 | 1741 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |