Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060728.3 | NP_493129.3 | 5 - 6 | 1192 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |