Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060728.3 | NP_493129.3 | 7 - 8 | 1422 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |