Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375235.2 | NP_001361848.1 | 9 - 10 | 1862 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |