Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eya-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025884.6 | NP_001021055.1 | 4 - 5 | 508 | 68 | |
NM_001381258.2 | NP_001367120.1 | 4 - 5 | 414 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |