Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383767.1 | 3 - 4 | 265 | 70 | ||
NM_061181.7 | NP_493582.3 | 3 - 4 | 265 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |