Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026695.4 | NP_001021866.1 | 4 - 5 | 816 | 62 | |
NM_061198.5 | NP_493599.2 | 3 - 4 | 679 | 62 | ||
NM_061199.6 | NP_493600.2 | 3 - 4 | 679 | 62 | ||
NM_061200.6 | NP_493601.2 | 3 - 4 | 674 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |