Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prp-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061345.10 | NP_493746.1 | 5 - 6 | 1210 | 65 | |
GenBank mRNA | U09415.1 | - | 5 - 6 | 1221 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |