Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prp-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061345.10 | NP_493746.1 | 7 - 8 | 1598 | 79 | |
GenBank mRNA | U09415.1 | - | 7 - 8 | 1609 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |