Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061349.8 | NP_493750.1 | 5 - 6 | 729 | 98 | |
GenBank mRNA | AJ252168.1 | - | 5 - 6 | 641 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |