Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061431.4 | NP_493832.1 | 2 - 3 | 166 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |