Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129167.3 | NP_001122639.1 | 2 - 3 | 106 | 82 | |
NM_061516.5 | NP_493917.2 | 2 - 3 | 100 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |