Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377691.1 | NP_001364697.1 | 2 - 3 | 315 | 96 | |
NM_001377692.2 | NP_001364548.1 | 2 - 3 | 321 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |