Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027043.3 | NP_001022214.1 | 1 - 2 | 224 | 70 | |
NM_001027044.4 | NP_001022215.1 | 1 - 2 | 228 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |