Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047270.4 | NP_001040735.2 | 5 - 6 | 1206 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |