Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061876.8 | NP_494277.3 | 9 - 10 | 1467 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |