Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361852.3 | NP_001348701.1 | 5 | 731 | 94 | |
NM_001361853.3 | NP_001348700.1 | 5 - 6 | 786 | 94 | ||
NM_001361854.3 | NP_001348702.1 | 5 | 731 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |