Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062169.6 | NP_494570.1 | 6 - 7 | 876 | 139 | |
NM_062170.5 | NP_494571.1 | 2 - 3 | 156 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |