Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062227.5 | NP_494628.2 | 1 - 2 | 81 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |