Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047259.5 | NP_001040724.1 | 17 - 18 | 3107 | 99 | |
NM_001047260.3 | NP_001040725.1 | 2 - 3 | 453 | 99 | ||
GenBank mRNA | AY314772.1 | - | 17 - 18 | 3090 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |