Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062447.9 | NP_494848.3 | 4 - 5 | 1429 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |