Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356833.4 | NP_001343753.1 | 2 | 417 | 94 | |
NM_001356834.3 | NP_001343752.1 | 2 - 3 | 192 | 94 | ||
NM_001377798.2 | NP_001364748.1 | 3 - 4 | 408 | 94 | ||
GenBank mRNA | AY585194.1 | - | 3 - 4 | 405 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |