Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062632.7 | NP_495033.2 | 8 - 9 | 1308 | 82 | |
GenBank mRNA | AF318613.2 | - | 8 - 9 | 1306 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |