Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027037.4 | NP_001022208.1 | 4 - 5 | 507 | 134 | |
NM_001027038.3 | NP_001022209.1 | 4 - 5 | 488 | 134 | ||
NM_001136345.4 | NP_001129817.1 | 4 - 5 | 502 | 134 | ||
NM_001347284.3 | NP_001334206.1 | 4 - 5 | 392 | 134 | ||
NR_144560.1 | 4 - 5 | 392 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |