Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lec-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062762.10 | NP_495163.1 | 6 - 7 | 874 | 153 | |
GenBank mRNA | AB038506.1 | - | 5 - 6 | 847 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |