Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062882.9 | NP_495283.2 | 2 - 3 | 532 | 68 | |
GenBank mRNA | DQ340628.1 | - | 2 - 3 | 495 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |