Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gld-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063078.4 | NP_495479.2 | 5 - 6 | 551 | 107 | |
NM_063079.7 | NP_495480.1 | 5 - 6 | 551 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |