Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026891.6 | NP_001022062.1 | 4 - 5 | 623 | 80 | |
NR_151558.1 | 3 - 4 | 582 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |