Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322736.1 | NP_001309489.1 | 2 - 3 | 438 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |