Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306541.3 | NP_001293470.1 | 2 - 3 | 863 | 75 | |
NM_001383885.1 | 6 - 7 | 1944 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |