Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063231.2 | NP_495632.1 | 4 - 5 | 772 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |