Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cki-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027138.6 | NP_001022309.1 | 3 - 4 | 544 | 101 | |
NM_001381510.2 | NP_001368240.1 | 2 - 3 | 415 | 101 | ||
NM_063241.7 | NP_495642.3 | 3 - 4 | 542 | 101 | ||
GenBank mRNA | AF179359.1 | - | 2 - 3 | 438 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |