Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063253.8 | NP_495654.1 | 5 - 6 | 2142 | 74 | |
GenBank mRNA | AF240692.1 | - | 5 - 6 | 2180 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |