Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063435.5 | NP_495836.2 | 2 - 3 | 296 | 83 | |
GenBank mRNA | AY275709.1 | - | 2 - 3 | 312 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |