Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sphk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026846.4 | NP_001022017.1 | 2 - 3 | 377 | 79 | |
NM_001267250.2 | NP_001254179.1 | 2 - 3 | 416 | 79 | ||
NM_001381535.1 | NP_001367075.1 | 2 - 3 | 226 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |