Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383908.1 | 8 - 9 | 990 | 169 | ||
GenBank mRNA | AJ745071.1 | - | 8 - 9 | 975 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |