Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063556.8 | NP_495957.1 | 10 - 11 | 1735 | 81 | |
GenBank mRNA | AY028165.1 | - | 10 - 11 | 1722 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |