Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026880.5 | NP_001022051.1 | 4 - 5 | 787 | 142 | |
NM_001383916.1 | 4 - 5 | 779 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |