Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-13A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063684.5 | NP_496085.3 | 6 - 7 | 1486 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |