Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063760.5 | NP_496161.1 | 3 - 4 | 391 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |