Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pro-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063823.5 | NP_496224.2 | 8 - 9 | 1678 | 85 | |
GenBank mRNA | EU068465.1 | - | 8 - 9 | 1677 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |