Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ect-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063917.6 | NP_496318.1 | 4 - 5 | 529 | 62 | |
NM_063918.3 | NP_496319.1 | 5 - 6 | 554 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |