Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063922.6 | NP_496323.1 | 3 - 4 | 799 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |