Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mix-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063930.8 | NP_496331.1 | 11 - 12 | 2900 | 114 | |
GenBank mRNA | U96387.1 | - | 11 - 12 | 2762 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |