Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063990.5 | NP_496391.1 | 4 - 5 | 713 | 65 | |
GenBank mRNA | AF008590.1 | - | 4 - 5 | 673 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |