Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026729.4 | NP_001021900.1 | 9 - 10 | 1349 | 90 | |
NM_001377875.3 | NP_001364585.1 | 9 - 10 | 1427 | 90 | ||
NM_001383953.1 | 8 - 9 | 1237 | 90 | |||
GenBank mRNA | AF173171.1 | - | 8 - 9 | 1259 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |