Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064035.9 | NP_496436.1 | 2 - 3 | 578 | 61 | |
GenBank mRNA | AB055111.1 | - | 2 - 3 | 600 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |